Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-fluoro-2-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. B45065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45065G 5 g
B450625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45065G Supplier TCI America Supplier No. B45065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 202865-72-3
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00042578
InChI Key QDCBSSUCEUMLGC-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoroiodobenzene, 4-fluoro-2-iodobromobenzene, 2-iodo-4-fluorobromobenzene, 1-bromo-4-fluoro-2-iodo-benzene, benzene, 1-bromo-4-fluoro-2-iodo, 2-bromo-5-fluoro-1-iodobenzene, pubchem3233, acmc-1cljm, ksc493k3j, 1-bromo-2-iodo-4-fluorobenzene
PubChem CID 2773370
IUPAC Name 1-bromo-4-fluoro-2-iodobenzene
SMILES C1=CC(=C(C=C1F)I)Br

Specifications

Color Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 300.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-fluoro-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.