accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFCl

CAS RN 202925-07-3

IUPAC Name: 2-chloro-1-fluoro-4-methoxybenzene
Synonyms: 2-chloro-1-fluoro-4-methoxybenzene
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: RWKKEILFRAYSDQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results