Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluoroanisole 97.0+%, TCI America™
SDP

Catalog No. C22175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22175G Supplier TCI America Supplier No. C22175G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 202925-07-3
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.572
MDL Number MFCD00070778
InChI Key RWKKEILFRAYSDQ-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoroanisole, benzene, 2-chloro-1-fluoro-4-methoxy, pubchem2660, acmc-1ckw3, 2-chloro-1-fluoro-4-methoxy-benzene, benzene,2-chloro-1-fluoro-4-methoxy, 2-chloranyl-1-fluoranyl-4-methoxy-benzene
PubChem CID 2773585
IUPAC Name 2-chloro-1-fluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)F)Cl

Specifications

Color Yellow
Boiling Point 189°C
Quantity 5 g
Formula Weight 160.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.