CAS RN 20302-14-1
IUPAC Name:
9,9-diphenyl-9H-fluorene
Synonyms:
9,9-diphenylfluorene
Molecular Weight (g/mol):
318.42
Molecular Formula:
C25H18
InChi Key:
BKQXUNGELBDWLS-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
1
–
1
of
1
results
1
9,9-Diphenylfluorene 98.0+%, TCI America™
CAS: 20302-14-1 Molecular Formula: C25H18 Molecular Weight (g/mol): 318.419 MDL Number: MFCD00093944 InChI Key: BKQXUNGELBDWLS-UHFFFAOYSA-N PubChem CID: 413569 IUPAC Name: 9,9-diphenylfluorene SMILES: C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=CC=C5