Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Diphenylfluorene 98.0+%, TCI America™
SDP

Catalog No. D35865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35865G 5 g
D358625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35865G Supplier TCI America Supplier No. D35865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20302-14-1
Molecular Formula C25H18
Molecular Weight (g/mol) 318.419
MDL Number MFCD00093944
InChI Key BKQXUNGELBDWLS-UHFFFAOYSA-N
PubChem CID 413569
IUPAC Name 9,9-diphenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=CC=C5

Specifications

Melting Point 226°C
Color White
Quantity 5 g
Formula Weight 318.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Diphenylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.