accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H2H2H

CAS RN 2031-90-5

IUPAC Name: (²H₁₂)pentane
Synonyms: (²H₁₂)pentane
Molecular Weight (g/mol): 84.22
Molecular Formula: C5H12
InChi Key: OFBQJSOFQDEBGM-HYVJACIRSA-N
SMILES: [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results