Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane-d{12}, 98%(Isotopic)

Catalog No. AA4234003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4234003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4234003 Supplier Thermo Scientific Chemicals Supplier No. 04234003
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2031-90-5 | C5H12 | 84.224 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2031-90-5
Molecular Formula C5H12
Molecular Weight (g/mol) 84.224
MDL Number MFCD00190495
InChI Key OFBQJSOFQDEBGM-HYVJACIRSA-N
Synonym pentane-d12, n-pentane-d12, 2 h?? pentane, pentane-d12, 98 atom % d
PubChem CID 137286
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,5-dodecadeuteriopentane
SMILES CCCCC

Specifications

Physical Form Liquid
Sensitivity Light
Quantity 1 g
Density 0.731 g/mL
Refractive Index 1.358
Flash Point −49°C (−56°F)
Melting Point -130°C
Boiling Point 36°C
Linear Formula CD3(CD2)3CD3
UN Number UN1265
Formula Weight 84.22
Percent Purity 98%
Chemical Name or Material n-Pentane-d{12}, Isotopic
Show More Show Less
Hazard Category H225-H304-H336
Hazard Statement GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES
EINECSNumber 203-692-4
TSCA Yes
storageNote1 2°C to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.