accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 203126-21-0

IUPAC Name: 1-methyl-4-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-methyl-4-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 218.19
Molecular Formula: C7H7F5S
InChi Key: MFSRNICSILXXMS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results