Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolylsulfur Pentafluoride 98.0+%, TCI America™
SDP

Catalog No. T28291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28291G Supplier TCI America Supplier No. T28291G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 203126-21-0
Molecular Formula C7H7F5S
Molecular Weight (g/mol) 218.19
MDL Number MFCD16817600
InChI Key MFSRNICSILXXMS-UHFFFAOYSA-N
Synonym 4-Methylphenylsulfur Pentafluoride
PubChem CID 50998489
IUPAC Name 1-methyl-4-(pentafluoro-λ⁶-sulfanyl)benzene
SMILES CC1=CC=C(C=C1)S(F)(F)(F)(F)F

Specifications

Color Colorless
Boiling Point 96°C
Quantity 1 g
UN Number 2810
Formula Weight 218.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolylsulfur Pentafluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.