accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2032-35-1

IUPAC Name: 2-bromo-1,1-diethoxyethane
Synonyms: 2-bromo-1,1-diethoxyethane
Molecular Weight (g/mol): 197.07
Molecular Formula: C6H13BrO2
InChi Key: LILXDMFJXYAKMK-UHFFFAOYSA-N
SMILES: CCOC(CBr)OCC
Boiling Point 
  • (6)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (7)
  • (6)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (9)
  • (3)