CAS RN 20327-23-5
IUPAC Name:
1-cyclopropylpiperazine
Synonyms:
1-cyclopropylpiperazine
Molecular Weight (g/mol):
126.2
Molecular Formula:
C7H14N2
InChi Key:
HNZJIWIXRPBFAN-UHFFFAOYSA-N
SMILES:
C1CC1N1CCNCC1
1
–
1
of
1
results
1
1-Cyclopropylpiperazine 98.0+%, TCI America™
CAS: 20327-23-5 Molecular Formula: C7H14N2 Molecular Weight (g/mol): 126.20 MDL Number: MFCD06254805 InChI Key: HNZJIWIXRPBFAN-UHFFFAOYSA-N PubChem CID: 4742004 IUPAC Name: 1-cyclopropylpiperazine SMILES: C1CC1N1CCNCC1