Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopropylpiperazine 98.0+%, TCI America™
SDP

Catalog No. C27941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27941G Supplier TCI America Supplier No. C27941G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20327-23-5
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.20
MDL Number MFCD06254805
InChI Key HNZJIWIXRPBFAN-UHFFFAOYSA-N
PubChem CID 4742004
IUPAC Name 1-cyclopropylpiperazine
SMILES C1CC1N1CCNCC1

Specifications

Color Yellow
Boiling Point 43°C
Quantity 1 g
UN Number 3267
Formula Weight 126.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Cyclopropylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.