accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNHBrBrN

CAS RN 2034-22-2

IUPAC Name: 2,4,5-tribromo-1H-imidazole
Synonyms: 2,4,5-tribromoimidazole
Molecular Weight (g/mol): 304.77
Molecular Formula: C3HBr3N2
InChi Key: JCGGPCDDFXIVQB-UHFFFAOYSA-N
SMILES: BrC1=NC(Br)=C(Br)N1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)