Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Tribromoimidazole 98.0+%, TCI America™
SDP

Catalog No. T22665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22665G 5 g
T226625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22665G Supplier TCI America Supplier No. T22665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2034-22-2
Molecular Formula C3HBr3N2
Molecular Weight (g/mol) 304.767
InChI Key JCGGPCDDFXIVQB-UHFFFAOYSA-N
Synonym 2,4,5-tribromoimidazole, 1h-imidazole, 2,4,5-tribromo, imidazole, 2,4,5-tribromo, 2,4,5-tribromoimidazo, tribromoimidazole, pubchem7150, 2,5-tribromoimidazole, acmc-1cmbx, timtec-bb sbb001252, imidazole,4,5-tribromo
PubChem CID 16253
IUPAC Name 2,4,5-tribromo-1H-imidazole
SMILES C1(=C(N=C(N1)Br)Br)Br

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 304.77
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Tribromoimidazole
RTECSNumber NI8660000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.