accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20348-23-6

OHNN

CAS RN 20348-23-6

IUPAC Name: 2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Synonyms: 2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: QQVXDMFULJVZLA-UHFFFAOYSA-N
SMILES: C1COC2=C(N1)N=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results