accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3C

CAS RN 20348-72-5

IUPAC Name: 1,2,3-trimethylcyclopentane
Synonyms: 1,2,3-trimethylcyclopentane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: VCWNHOPGKQCXIQ-UHFFFAOYNA-N
SMILES: CC1CCC(C)C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results