accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3

CAS RN 2035-99-6

IUPAC Name: 3-methylbutyl octanoate
Synonyms: isoamyl octanoate
Molecular Weight (g/mol): 214.35
Molecular Formula: C13H26O2
InChi Key: XKWSWANXMRXDES-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results