Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl n-Octanoate (contains 2-Methylbutyl n-Octanoate), TCI America™
SDP

Catalog No. O003125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O003125ML Supplier TCI America Supplier No. O003125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2035-99-6
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.35
MDL Number MFCD00048917
InChI Key XKWSWANXMRXDES-UHFFFAOYSA-N
Synonym Isopentyl n-Octanoate, n-Octanoic Acid Isoamyl Ester
PubChem CID 16255
ChEBI CHEBI:87536
IUPAC Name 3-methylbutyl octanoate
SMILES CCCCCCCC(=O)OCCC(C)C

Specifications

Color Yellow
Boiling Point 250°C
Quantity 25 mL
Formula Weight 214.35
Physical Form Liquid
Chemical Name or Material Isoamyl n-Octanoate (contains 2-Methylbutyl n-Octanoate)
EINECSNumber (2)-0782
RTECSNumber RH0770000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.