accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHOHOH2NOHNOHOHHCl

CAS RN 203515-86-0

IUPAC Name: 5-amino-N1,N3-bis(2,3-dihydroxypropyl)benzene-1,3-dicarboxamide hydrochloride
Synonyms: 5-amino-N1,N3-bis(2,3-dihydroxypropyl)benzene-1,3-dicarboxamide hydrochloride
Molecular Weight (g/mol): 363.8
Molecular Formula: C14H22ClN3O6
InChi Key: GNBBCFFLJFKAHB-UHFFFAOYNA-N
SMILES: Cl.NC1=CC(=CC(=C1)C(=O)NCC(O)CO)C(=O)NCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results