Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-N,N'-bis(2,3-dihydroxypropyl)isophthalamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A19515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19515G Supplier TCI America Supplier No. A19515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 203515-86-0
Color White-Yellow
Molecular Formula C14H22ClN3O6
MDL Number MFCD00270835
Quantity 5 g
InChI Key GNBBCFFLJFKAHB-UHFFFAOYNA-N
SMILES Cl.NC1=CC(=CC(=C1)C(=O)NCC(O)CO)C(=O)NCC(O)CO
IUPAC Name 5-amino-N1,N3-bis(2,3-dihydroxypropyl)benzene-1,3-dicarboxamide hydrochloride
Molecular Weight (g/mol) 363.80
PubChem CID 44629992
Formula Weight 363.80
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-N,N′-bis(2,3-dihydroxypropyl)isophthalamide Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.