accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 203626-45-3

IUPAC Name: 2-(acetyloxy)-5-cyanophenyl acetate
Synonyms: 2-(acetyloxy)-5-cyanophenyl acetate
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: OYRIDYHXQXSNCH-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(OC(C)=O)C=C(C=C1)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results