CAS RN 203866-17-5
IUPAC Name:
1-tert-butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate
Synonyms:
1-tert-butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol):
265.26
Molecular Formula:
C11H17F2NO4
InChi Key:
RQDZKOOUQIDZOG-UHFFFAOYNA-N
SMILES:
COC(=O)C1CC(F)(F)CN1C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Sigma Aldrich N-Boc-Trans-4-Fluoro-L-Proline Methyl Ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.