accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2039-82-9

IUPAC Name: 1-bromo-4-ethenylbenzene
Synonyms: 4-bromostyrene
Molecular Weight (g/mol): 183.05
Molecular Formula: C8H7Br
InChi Key: WGGLDBIZIQMEGH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C)C=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)