accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNSOONHClHCl

CAS RN 203911-27-7

IUPAC Name: 5-(1,4-diazepane-1-sulfonyl)isoquinoline dihydrochloride
Synonyms: fasudil dihydrochloride
Molecular Weight (g/mol): 364.29
Molecular Formula: C14H19Cl2N3O2S
InChi Key: NOXXIYDYFSNHDF-UHFFFAOYSA-N
SMILES: Cl.Cl.O=S(=O)(N1CCCNCC1)C1=C2C=CN=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results