accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20405-64-5

IUPAC Name: (cuprioselanyl)copper
Synonyms: copper selenide
Molecular Weight (g/mol): 206.06
Molecular Formula: Cu2Se
InChi Key: KTLOQXXVQYUCJU-UHFFFAOYSA-N
SMILES: [Cu][Se][Cu]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)