Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) selenide

Catalog No. 4816295G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-162-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-162-95G Supplier Sigma Aldrich Supplier No. 4816295G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Copper(I) selenide (Cu2Se) is a semiconductor material, which exists in cubic, tetragonal or orthorhombic forms. It forms thin films, which can be used as solar cell materials, optical filters, thermoelectric converters, electro-optical devices, and microwave shield coatings.

Specifications

CAS 20405-64-5
Density 6.84 g/mL (at 25°C)
Molecular Formula Cu2Se
Linear Formula Cu2Se
MDL Number MFCD00049908
Quantity 5 g
Synonym Cuprous selenide; Dicopper monoselenide; Dicopper selenide
Molecular Weight (g/mol) 206.05
Percent Purity 99.95% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.