accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(E)

CAS RN 20407-84-5

IUPAC Name: (2E)-dodec-2-enal
Synonyms: 2-dodecenal
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: SSNZFFBDIMUILS-ZHACJKMWSA-N
SMILES: CCCCCCCCC\C=C\C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results