Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Dodecenal 90.0+%, TCI America™
SDP

Catalog No. D206825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D206825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D206825ML Supplier TCI America Supplier No. D206825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20407-84-5
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00014674
InChI Key SSNZFFBDIMUILS-ZHACJKMWSA-N
PubChem CID 5283361
IUPAC Name (E)-dodec-2-enal
SMILES CCCCCCCCCC=CC=O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 mL
Formula Weight 182.31
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Dodecenal
RTECSNumber JR5150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.