accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNN

CAS RN 204078-93-3

IUPAC Name: 2-(4-methyl-1,4-diazepan-1-yl)benzonitrile
Synonyms: 2-(4-methyl-1,4-diazepan-1-yl)benzonitrile
Molecular Weight (g/mol): 215.3
Molecular Formula: C13H17N3
InChi Key: WKJLOVRBRYDGGK-UHFFFAOYSA-N
SMILES: CN1CCCN(CC1)C1=CC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results