Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(4-Methylperhydro-1,4-diazepin-1-yl)benzonitrile, 95%, Thermo Scientific™
Specifications
Specifications
| CAS | 204078-93-3 |
| CAS Min % | 94.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C13H17N3 |
| Quantity | 250mg |
| MDL Number | MFCD09065002 |
| Synonym | 2-4-methyl-1,4-diazepan-1-yl benzonitrile, 2-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 2-4-methyl-1-homopiperazinyl benzonitrile, 2-4-methylhomopiperazin-1-yl benzonitrile, 2-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile |
| InChI Key | WKJLOVRBRYDGGK-UHFFFAOYSA-N |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?