accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2043-00-7

IUPAC Name: 1-(2-phenylethenyl)naphthalene
Synonyms: 1-(2-phenylethenyl)naphthalene
Molecular Weight (g/mol): 230.31
Molecular Formula: C18H14
InChi Key: QAVDMWIHZMXKFR-UHFFFAOYSA-N
SMILES: C(=CC1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results