Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Styrylnaphthalene 95.0+%, TCI America™
SDP

Catalog No. S04881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04881G Supplier TCI America Supplier No. S04881G

Chemical Identifiers

CAS 2043-00-7
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00021574
InChI Key QAVDMWIHZMXKFR-BUHFOSPRSA-N
PubChem CID 1549709
IUPAC Name 1-[(E)-2-phenylethenyl]naphthalene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC3=CC=CC=C32

Specifications

Color Yellow
Quantity 1 g
Formula Weight 230.31
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Styrylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.