accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2043-54-1

FFFFFFFFFFFFFFFFFFFFFI

CAS RN 2043-54-1

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-12-iodododecane
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-12-iodododecane
Molecular Weight (g/mol): 674.04
Molecular Formula: C12H4F21I
InChi Key: HVWXRMINOYZYCK-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results