accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2043361-32-4

BFFFFCH3CH3H3CCH3NNFCH3H3CCH3CH3(?)(?)

CAS RN 2043361-32-4

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2-fluoro-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-2-fluoro-1λ⁵-imidazol-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 494.4
Molecular Formula: C27H36BF5N2
InChi Key: ZXIKPDSPOFQTLH-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1F)C1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)