Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetra-N-Butylammonium Decatungstate

Catalog No. 900432250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-043-2250MG 250 mg
90-043-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-043-2250MG Supplier Sigma Aldrich Supplier No. 900432250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

When used in conjunction with cobaloxime pyridine chloride (341630), this complex can effect the catalytic, room-temperature dehydrogenation of unactivated alkanes under solar irradiation with hydrogen as the sole byproduct.Acceptorless dehydrogenation of small molecules through cooperative base metal catalysisC(sp3)-H functionalizations of light hydrocarbons using decatungstate photocatalysis in flow

Specifications

CAS 2043361-32-4
Molecular Formula C64H144N4O32W10
Linear Formula C64H144N4O32W10
Quantity 250 mg
Synonym 1-Butanaminium,-»N,N,N-tributyl-, eicosa-mu-oxodi-mu5-oxodecaoxodecatungstate (4:1); 1-Butanaminium,-»N,N,N-tributyl-, salt with tungstic acid (4:1); TBADT; Tetrakis(N,N,N-tributyl-1-butanaminium) tungstate; Tetrakis(tetrabutylammonium) decatungstate
Molecular Weight (g/mol) 3320.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.