accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2044-21-5

IUPAC Name: dinonylamine
Synonyms: dinonylamine
Molecular Weight (g/mol): 269.52
Molecular Formula: C18H39N
InChi Key: MFHKEJIIHDNPQE-UHFFFAOYSA-N
SMILES: CCCCCCCCCNCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results