Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dinonylamine 97.0+%, TCI America™
SDP

Catalog No. D26765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26765G Supplier TCI America Supplier No. D26765G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2044-21-5
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00027338
InChI Key MFHKEJIIHDNPQE-UHFFFAOYSA-N
PubChem CID 94761
IUPAC Name N-nonylnonan-1-amine
SMILES CCCCCCCCCNCCCCCCCCC

Specifications

Color White
Quantity 5 g
UN Number 3259
Formula Weight 269.52
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dinonylamine
EINECSNumber (2)-0138
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.