accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3

CAS RN 2044-22-6

IUPAC Name: trinonylamine
Synonyms: amines, tri-C8-10-alkyl
Molecular Weight (g/mol): 395.76
Molecular Formula: C27H57N
InChi Key: ZQJAONQEOXOVNR-UHFFFAOYSA-N
SMILES: CCCCCCCCCN(CCCCCCCCC)CCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results