Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trinonylamine 98.0+%, TCI America™
SDP

Catalog No. T18265ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18265ML Supplier TCI America Supplier No. T18265ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2044-22-6
Molecular Formula C27H57N
Molecular Weight (g/mol) 395.76
MDL Number MFCD00053817
InChI Key ZQJAONQEOXOVNR-UHFFFAOYSA-N
PubChem CID 137295
IUPAC Name N,N-di(nonyl)nonan-1-amine
SMILES CCCCCCCCCN(CCCCCCCCC)CCCCCCCCC

Specifications

Color Yellow
Boiling Point 270°C
Quantity 5 mL
Formula Weight 395.76
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trinonylamine
EINECSNumber (2)-0139
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.