accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OCH3O

CAS RN 2044-64-6

IUPAC Name: N,N-dimethyl-3-oxobutanamide
Synonyms: N,N-dimethyl-3-oxobutanamide
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: YPEWWOUWRRQBAX-UHFFFAOYSA-N
SMILES: CN(C)C(=O)CC(C)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results