Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylacetoacetamide solution

Catalog No. 537373500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-737-3500ML 500 mL
53-737-32L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-737-3500ML Supplier Sigma Aldrich Supplier No. 537373500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is an 80% solution of N,N-dimethylacetoacetamide (DMAA) in water.

Specifications

CAS 2044-64-6
Density 1.067 g/mL (at 25°C)
Boiling Point 105°C
Molecular Formula C6H11NO2
Refractive Index n20/D 1.447
Linear Formula CH3COCH2CON(CH3)2
MDL Number MFCD00038243
Quantity 500 mL
Synonym DMAA; N,N-Dimethyl-3-oxobutyramide
Molecular Weight (g/mol) 129.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.