accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20440-93-1

IUPAC Name: 4-nitro-N,N-bis(4-nitrophenyl)aniline
Synonyms: 4-nitro-N,N-bis(4-nitrophenyl)aniline
Molecular Weight (g/mol): 380.32
Molecular Formula: C18H12N4O6
InChi Key: LSNJBIDKQIRWRQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N(C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results