Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-nitrophenyl)amine 96.0+%, TCI America™
SDP

Catalog No. T23335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23335G 5 g
T233325G 25 g
T23331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T23335G Supplier TCI America Supplier No. T23335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20440-93-1
Molecular Formula C18H12N4O6
Molecular Weight (g/mol) 380.316
MDL Number MFCD00024636
InChI Key LSNJBIDKQIRWRQ-UHFFFAOYSA-N
PubChem CID 88538
IUPAC Name 4-nitro-N,N-bis(4-nitrophenyl)aniline
SMILES C1=CC(=CC=C1N(C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 280°C
Quantity 5 g
Formula Weight 380.32
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-nitrophenyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.