accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClBr

CAS RN 20443-99-6

IUPAC Name: 1-(bromomethyl)-2,4-dichlorobenzene
Synonyms: 1-(bromomethyl)-2,4-dichlorobenzene
Molecular Weight (g/mol): 239.92
Molecular Formula: C7H5BrCl2
InChi Key: RGLQSFFFIREZFV-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(CBr)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results