Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D44755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44755G 5 g
D44751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44755G Supplier TCI America Supplier No. D44755G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20443-99-6
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.92
MDL Number MFCD03208523
InChI Key RGLQSFFFIREZFV-UHFFFAOYSA-N
PubChem CID 10988373
IUPAC Name 1-(bromomethyl)-2,4-dichlorobenzene
SMILES ClC1=CC(Cl)=C(CBr)C=C1

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 112°C
Quantity 5 g
UN Number 3261
Formula Weight 239.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichlorobenzyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.