accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2045-00-3

IUPAC Name: (2-aminophenyl)arsonic acid
Synonyms: o-aminobenzenearsonic acid
Molecular Weight (g/mol): 217.06
Molecular Formula: C6H8AsNO3
InChi Key: LQCOCUQCZYAYQK-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1[As](O)(O)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)