Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Arsanilic Acid 98%, Thermo Scientific™

Catalog No. AC160620500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC160620500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC160620500 Supplier Thermo Scientific Chemicals Supplier No. 160620500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2045-00-3
Molecular Formula C6H8AsNO3
Molecular Weight (g/mol) 217.056
InChI Key LQCOCUQCZYAYQK-UHFFFAOYSA-N
Synonym o-arsanilic acid, 2-aminobenzenearsonic acid, o-aminobenzenearsonic acid, 2-aminophenyl arsonic acid, arsenical, unii-4p7t2ph7dl, 2-arsanilic acid, o-aminophenylarsonic acid, 4p7t2ph7dl, arsonic acid,as-2-aminophenyl
PubChem CID 73161
IUPAC Name (2-aminophenyl)arsonic acid
SMILES C1=CC=C(C(=C1)N)[As](=O)(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 145°C to 150°C
Color Brown to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NC6H4ASO3H2
Quantity 50 g
Beilstein 16, I, 463
Formula Weight 217.06
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material o-Arsanilic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.