CAS RN 2046-18-6
CAS RN 2046-18-6
IUPAC Name:
4-phenylbutanenitrile
Synonyms:
G-phenylpropylcyanide
Molecular Weight (g/mol):
145.21
Molecular Formula:
C10H11N
InChi Key:
ICMVGKQFVMTRLB-UHFFFAOYSA-N
SMILES:
N#CCCCC1=CC=CC=C1
1
–
1
of
1
results
1
4-Phenylbutyronitrile 98.0+%, TCI America™

CAS: 2046-18-6 Molecular Formula: C10H11N Molecular Weight (g/mol): 145.21 MDL Number: MFCD00001973 InChI Key: ICMVGKQFVMTRLB-UHFFFAOYSA-N Synonym: 4-phenylbutyronitrile,benzenebutanenitrile,3-cyanopropyl benzene,butyronitrile, 4-phenyl,g-phenylpropylcyanide,4-phenyl-butyronitrile,acmc-209fai,gamma-phenylbutyronitrile,1-cyano-3-phenylpropane,d0ft2z PubChem CID: 74897 IUPAC Name: 4-phenylbutanenitrile SMILES: N#CCCCC1=CC=CC=C1