Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylbutyronitrile 98.0+%, TCI America™
SDP

Catalog No. P102525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P102525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P102525ML Supplier TCI America Supplier No. P102525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2046-18-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.21
MDL Number MFCD00001973
InChI Key ICMVGKQFVMTRLB-UHFFFAOYSA-N
Synonym 4-phenylbutyronitrile, benzenebutanenitrile, 3-cyanopropyl benzene, butyronitrile, 4-phenyl, g-phenylpropylcyanide, 4-phenyl-butyronitrile, acmc-209fai, gamma-phenylbutyronitrile, 1-cyano-3-phenylpropane, d0ft2z
PubChem CID 74897
IUPAC Name 4-phenylbutanenitrile
SMILES N#CCCCC1=CC=CC=C1

Specifications

Boiling Point 130°C
Quantity 25 mL
UN Number 3276
Formula Weight 145.21
Percent Purity ≥98.0% (GC)
Grade ACS
Physical Form Liquid
Chemical Name or Material 4-Phenylbutyronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.