CAS RN 2047-91-8
IUPAC Name:
1H,2H,3H,4H-cyclopenta[b]indole
Synonyms:
1H,2H,3H,4H-cyclopenta[b]indole
Molecular Weight (g/mol):
157.22
Molecular Formula:
C11H11N
InChi Key:
HZDXFZHFEASSBM-UHFFFAOYSA-N
SMILES:
C1CC2=C(C1)C1=CC=CC=C1N2
1
–
1
of
1
results
1
1,2,3,4-Tetrahydrocyclopenta[b]indole 97.0+%, TCI America™
CAS: 2047-91-8 Molecular Formula: C11H11N Molecular Weight (g/mol): 157.216 MDL Number: MFCD00225387 InChI Key: HZDXFZHFEASSBM-UHFFFAOYSA-N PubChem CID: 270305 IUPAC Name: 1,2,3,4-tetrahydrocyclopenta[b]indole SMILES: C1CC2=C(C1)NC3=CC=CC=C23